3-Fluoro-4-nitroaniline ≥95.0% (by GC)

Supplier: TCI America

F0787-1G F0787-5G
TCF0787-1GEA 61.62 CAD
TCF0787-1G TCF0787-5G
3-Fluoro-4-nitroaniline ≥95.0% (by GC)
3-Fluoro-4-nitroaniline

CAS Number: 2369-13-3
MDL Number: MFCD03412239
Molecular Formula: C6H5FN2O2
Molecular Weight: 156.12
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 161

Formula: C₆H₅FN₂O₂
Melting Pt: 161 °C
MDL Number: MFCD03412239
CAS Number: 2369-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51dd51cddfda073fded68ca7fa365dee0affaebb&vid=4a4a5367603778782d3bde5a7679b20d2c000f0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR