2-Fluoro-5-iodobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

F0786-1G F0786-5G
TCF0786-1GEA 89.27 CAD
TCF0786-1G TCF0786-5G
2-Fluoro-5-iodobenzoic acid ≥98.0% (by titrimetric analysis)
2-Fluoro-5-iodobenzoic acid

CAS Number: 124700-41-0
MDL Number: MFCD03094517
Molecular Formula: C7H4FIO2
Molecular Weight: 266.01
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 165

Formula: C₇H₄FIO₂
Melting Pt: 165 °C
MDL Number: MFCD03094517
CAS Number: 124700-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bba761334d240bb4607c7e94b5eff17d8a26494&vid=4bba761334d240bb4607c7e94b5eff17d8a26494&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR