2-Fluoro-4-biphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

F0785-1G F0785-5G
TCF0785-1GEA 60.04 CAD
TCF0785-1G TCF0785-5G
2-Fluoro-4-biphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-4-biphenylboronic acid

CAS Number: 178305-99-2
MDL Number: MFCD01075707
Molecular Formula: C12H10BFO2
Molecular Weight: 216.02
Form: Crystal
Color: White
Melting point (°C): 248


Other Product Names / Synonyms: 3-Fluoro-4-phenylbenzeneboronic Acid

Formula: C₁₂H₁₀BFO₂
Melting Pt: 248 °C
MDL Number: MFCD01075707
CAS Number: 178305-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=153176d7121993e48eaada83b893a20623290d36&vid=153176d7121993e48eaada83b893a20623290d36&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR