Fluorescein isothiocyanate (mixture of 5 and 6 isomers) ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

F0784-100MG F0784-1G
TCF0784-100MGEA 224.36 CAD
TCF0784-100MG TCF0784-1G
Fluorescein isothiocyanate (mixture of 5 and 6 isomers) ≥97.0% (by HPLC, titration analysis)
Fluorescein isothiocyanate (mixture of 5 and 6 isomers)

CAS Number: 27072-45-3
MDL Number: MFCD00005063
Molecular Formula: C21H11NO5S
Molecular Weight: 389.38
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Stains & Dyes, Fluorometry & Fluorescent Imaging, Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Isothiocyanatofluorescein

Formula: C₂₁H₁₁NO₅S
MDL Number: MFCD00005063
CAS Number: 27072-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cce5ceeb117e3aa7443b5d2ba2ead266ea24c6e1&vid=cce5ceeb117e3aa7443b5d2ba2ead266ea24c6e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR