2-(2-Fluorophenyl)ethanol ≥95.0% (by GC)

Supplier: TCI America

F0778-25G F0778-5G
TCF0778-25GEA 195.92 CAD
TCF0778-25G TCF0778-5G
2-(2-Fluorophenyl)ethanol ≥95.0% (by GC)
2-(2-Fluorophenyl)ethanol

CAS Number: 50919-06-7
MDL Number: MFCD00002887
Molecular Formula: C8H9FO
Molecular Weight: 140.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 98
Specific Gravity (20/20): 1.13


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluorophenethyl Alcohol, 2-(2-Fluorophenyl)ethyl Alcohol

Formula: C₈H₉FO
Boiling Pt: 137 °C
Density: 1.13 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00002887
CAS Number: 50919-06-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35ba864cc2c6321bb724a8873e128b34da45bf42&vid=35ba864cc2c6321bb724a8873e128b34da45bf42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR