4'-Fluoro-3'-nitroacetophenone ≥98.0% (by GC)

Supplier: TCI America

F0777-25G F0777-5G
TCF0777-25GEA 437.66 CAD
TCF0777-25G TCF0777-5G
4'-Fluoro-3'-nitroacetophenone ≥98.0% (by GC)
4'-Fluoro-3'-nitroacetophenone

CAS Number: 400-93-1
MDL Number: MFCD00115369
Molecular Formula: C8H6FNO3
Molecular Weight: 183.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 50

Formula: C₈H₆FNO₃
Boiling Pt: 120 °C
Melting Pt: 50 °C
MDL Number: MFCD00115369
CAS Number: 400-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a0cc0925044f99d13947b5e78a9117da8919bbc&vid=9a0cc0925044f99d13947b5e78a9117da8919bbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR