2-(3-Fluorophenyl)ethanol ≥98.0% (by GC)

Supplier: TCI America

F0776-1G F0776-5G
TCF0776-1GEA 58.46 CAD
TCF0776-1G TCF0776-5G
2-(3-Fluorophenyl)ethanol ≥98.0% (by GC)
2-(3-Fluorophenyl)ethanol

CAS Number: 52059-53-7
MDL Number: MFCD00045998
Molecular Formula: C8H9FO
Molecular Weight: 140.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 107
Specific Gravity (20/20): 1.13


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(3-Fluorophenyl)ethyl Alcohol, 3-Fluorophenethyl Alcohol

Formula: C₈H₉FO
Boiling Pt: 90 °C
Density: 1.13 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00045998
CAS Number: 52059-53-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98b5455b273c94f0ed44c7f00a403a152fd997a2&vid=98b5455b273c94f0ed44c7f00a403a152fd997a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR