5-Fluoro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 5-Fluoro-2-methoxybenzeneboronic acid

F0775-25G F0775-5G
TCF0775-25GEA 436.08 CAD
TCF0775-25G TCF0775-5G
5-Fluoro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
5-Fluoro-2-methoxyphenylboronic acid

CAS Number: 179897-94-0
MDL Number: MFCD01863526
Molecular Formula: C7H8BFO3
Molecular Weight: 169.95
Form: Crystal
Color: White
Melting point (°C): 158


Other Product Names / Synonyms: 5-Fluoro-2-methoxybenzeneboronic Acid

Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Melting Pt: 158 °C
MDL Number: MFCD01863526
CAS Number: 179897-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e36c361d2d745aae54254cdea4616ee020e5de3&vid=8e36c361d2d745aae54254cdea4616ee020e5de3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR