3-Fluoro-5-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America

F0774-1G F0774-5G
TCF0774-1GEA 75.84 CAD
TCF0774-1G TCF0774-5G
3-Fluoro-5-(trifluoromethyl)benzaldehyde ≥98.0% (by GC)
3-Fluoro-5-(trifluoromethyl)benzaldehyde

CAS Number: 188815-30-7
MDL Number: MFCD00061312
Molecular Formula: C8H4F4O
Molecular Weight: 192.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 71
Specific Gravity (20/20): 1.41

Formula: C₈H₄F₄O
Boiling Pt: 175 °C
Density: 1.41 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00061312
CAS Number: 188815-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb34180b8fcb63f0a80754de35f162fff1ed85ca&vid=eb34180b8fcb63f0a80754de35f162fff1ed85ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR