2-Fluoro-4-iodopyridine ≥98.0% (by GC)

Supplier: TCI America

F0773-1G F0773-5G
TCF0773-1GEA 63.2 CAD
TCF0773-1G TCF0773-5G
2-Fluoro-4-iodopyridine ≥98.0% (by GC)
2-Fluoro-4-iodopyridine

CAS Number: 22282-70-8
MDL Number: MFCD03092926
Molecular Formula: C5H3FIN
Molecular Weight: 222.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 58

Formula: C₅H₃FIN
MW: 222.99 g/mol
Melting Pt: 58 °C
MDL Number: MFCD03092926
CAS Number: 22282-70-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8ab9a81f768798a42a2bd04a0449da70185f505&vid=a8ab9a81f768798a42a2bd04a0449da70185f505&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR