(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid ≥98.0%

Supplier: TCI America

Synonyms: O-tert-Butyl-Nalpha-Fmoc-D-tyrosine, Nalpha-Fmoc-O-tert-butyl-D-tyrosine, Fmoc-D-Tyr(tBu)-OH, N-Fmoc-O-tert-butyl-D-tyrosine

F0772-1G F0772-5G
TCF0772-1GEA 82.16 CAD
TCF0772-1G TCF0772-5G
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid ≥98.0%
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid

CAS Number: 118488-18-9
MDL Number: MFCD00065684
Molecular Formula: C28H29NO5
Molecular Weight: 459.54
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 152
Specific rotation [a]20/D: 30 deg (C=1, DMF)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: O-tert-Butyl-Nalpha-Fmoc-D-tyrosine, Fmoc-D-Tyr(tBu)-OH, Nalpha-Fmoc-O-tert-butyl-D-tyrosine

Formula: C28H29NO5
MW: 459.5 g/mol
Melting Pt: 152 °C
MDL Number: MFCD00065684
CAS Number: 118488-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ea7992f83ea2e15fd5f46d403a5b039f1719e01&vid=6ea7992f83ea2e15fd5f46d403a5b039f1719e01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR