2-Fluoro-5-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 6-Fluoropyridine-3-boronic acid, 2-Fluoropyridine-5-boronic acid, 2-Fluoro-5-pyridylboronic acid, 2-Fluoro-5-pyridinylboronic acid, 6-Fluoro-3-pyridinylboronic acid

F0770-1G F0770-5G
TCF0770-1GEA 84.37 CAD
TCF0770-1G TCF0770-5G
2-Fluoro-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Fluoro-5-pyridineboronic acid

CAS Number: 351019-18-6
MDL Number: MFCD03411559
Molecular Formula: C5H5BFNO2
Molecular Weight: 140.91
Form: Crystal
Color: White
Melting point (°C): 172


Other Product Names / Synonyms: 6-Fluoro-3-pyridylboronic Acid

Formula: C₅H₅BFNO₂
MW: 140.91
Melting Pt: 172 °C
MDL Number: MFCD03411559
CAS Number: 351019-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbf45f2db722dad2fd29cc437b6083dfa1531a22&vid=bbf45f2db722dad2fd29cc437b6083dfa1531a22&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR