α,α,α,4-Tetrafluoro-m-toluic acid ≥98.0%

Supplier: TCI America

F0760-1G F0760-5G
TCF0760-1GEA 28.44 CAD
TCF0760-1G TCF0760-5G
α,α,α,4-Tetrafluoro-m-toluic acid ≥98.0%
α,α,α,4-Tetrafluoro-m-toluic acid

CAS Number: 67515-55-3
MDL Number: MFCD00061294
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 115


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,4-Tetrafluoro-m-toluic Acid

Formula: C₈H₄F₄O₂
MW: 208.11
Melting Pt: 115 °C
MDL Number: MFCD00061294
CAS Number: 67515-55-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24f85dcfd831dbb3aca74e4ab51536792d25900e&vid=24f85dcfd831dbb3aca74e4ab51536792d25900e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR