α,α,α,5-Tetrafluoro-m-toluic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-Fluoro-5-trifluoromethylbenzoic acid, 3-Fluoro-5-(trifluoromethyl)benzoic acid

F0759-1G F0759-5G
TCF0759-1GEA 51.19 CAD
TCF0759-1G TCF0759-5G
α,α,α,5-Tetrafluoro-m-toluic acid ≥98.0%
α,α,α,5-Tetrafluoro-m-toluic acid

CAS Number: 161622-05-5
MDL Number: MFCD00061293
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 104


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,5-Tetrafluoro-m-toluic Acid

Formula: C₈H₄F₄O₂
MW: 208.11
Melting Pt: 104 °C
MDL Number: MFCD00061293
CAS Number: 161622-05-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18e095dcffad17b6480ac37551026a8de28f4dbd&vid=18e095dcffad17b6480ac37551026a8de28f4dbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR