α,α,α,3-Tetrafluoro-p-toluic acid ≥98.0%
Supplier: TCI America
F0758-1G
F0758-5G
TCF0758-1GEA
57.83
CAD
TCF0758-1G
TCF0758-5G
α,α,α,3-Tetrafluoro-p-toluic acid ≥98.0%
α,α,α,3-Tetrafluoro-p-toluic acid
CAS Number: 115754-21-7
MDL Number: MFCD00236279
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 176
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,3-Tetrafluoro-p-toluic Acid
Formula:
C₈H₄F₄O₂ MW: 208.11 |
MDL Number:
MFCD00236279 CAS Number: 115754-21-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...