α,α,α,6-Tetrafluoro-o-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0757-1G F0757-5G
TCF0757-1GEA 61.15 CAD
TCF0757-1G TCF0757-5G
α,α,α,6-Tetrafluoro-o-toluic acid ≥98.0% (by GC, titration analysis)
α,α,α,6-Tetrafluoro-o-toluic acid

CAS Number: 32890-94-1
MDL Number: MFCD00040981
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 88


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,6-Tetrafluoro-o-toluic Acid

Formula: C₈H₄F₄O₂
Melting Pt: 88 °C
MDL Number: MFCD00040981
CAS Number: 32890-94-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e403e95871dc74fb39f5128bef54bcf9e95756fc&vid=e403e95871dc74fb39f5128bef54bcf9e95756fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR