N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine ≥98.0%

Supplier: TCI America

Synonyms: Nα-Fmoc-S-trityl-D-cysteine, Fmoc-D-Cys(Trt)-OH, N-Fmoc-S-trityl-D-cysteine

F0752-1G F0752-5G
TCF0752-1GEA 50.56 CAD
TCF0752-1G TCF0752-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine ≥98.0%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine

CAS Number: 167015-11-4
MDL Number: MFCD00151922
Molecular Formula: C37H31NO4S
Molecular Weight: 585.72
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 174
Specific rotation [a]20/D: -18 deg (C=1, THF)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-S-trityl-D-cysteine, Fmoc-D-Cys(Trt)-OH

Formula: C₃₇H₃₁NO₄S
MW: 585.72 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00151922
CAS Number: 167015-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=781c4cd5d74b60a76c70d015d138565f864cf103&vid=781c4cd5d74b60a76c70d015d138565f864cf103&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR