5-Fluoro-2-methylbenzaldehyde ≥95.0% (by GC)

Supplier: TCI America

F0746-25G F0746-5G
TCF0746-25GEA 635.16 CAD
TCF0746-25G TCF0746-5G
5-Fluoro-2-methylbenzaldehyde ≥95.0% (by GC)
5-Fluoro-2-methylbenzaldehyde

CAS Number: 22062-53-9
MDL Number: MFCD02261763
Molecular Formula: C8H7FO
Molecular Weight: 138.14
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 77
Specific Gravity (20/20): 1.16

Formula: C₈H₇FO
Boiling Pt: 99 °C
Density: 1.16 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD02261763
CAS Number: 22062-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5030681456058dddccaa90228bc25d3cdf290f4&vid=c5030681456058dddccaa90228bc25d3cdf290f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR