3-(4-Fluorophenyl)-1-isopropylindole ≥98.0% (by GC)

Supplier: TCI America

F0743-25G F0743-5G
TCF0743-25GEA 84.37 CAD
TCF0743-25G TCF0743-5G
3-(4-Fluorophenyl)-1-isopropylindole ≥98.0% (by GC)
3-(4-Fluorophenyl)-1-isopropylindole

CAS Number: 93957-49-4
MDL Number: MFCD01075733
Molecular Formula: C17H16FN
Molecular Weight: 253.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 97

Formula: C₁₇H₁₆FN
Melting Pt: 97 °C
MDL Number: MFCD01075733
CAS Number: 93957-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7862cb89755f57f8016df6dce5daa26bb355af2a&vid=7862cb89755f57f8016df6dce5daa26bb355af2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR