2-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
F0739-25G
F0739-5G
TCF0739-25GEA
641.48
CAD
TCF0739-25G
TCF0739-5G
2-Fluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
2-Fluoro-3-pyridineboronic acid
CAS Number: 174669-73-9
MDL Number: MFCD03411557
Molecular Formula: C5H5BFNO2
Molecular Weight: 140.91
Form: Crystal
Color: White
Melting point (°C): 172
Other Product Names / Synonyms: 2-Fluoro-3-pyridylboronic Acid
Formula:
C₅H₅BFNO₂ Melting Pt: 172 °C |
MDL Number:
MFCD03411557 CAS Number: 174669-73-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...