4-Fluoro-2-nitrobenzoic acid ≥98.0%

Supplier: TCI America

F0737-1G F0737-5G
TCF0737-1GEA 83.74 CAD
TCF0737-1G TCF0737-5G
4-Fluoro-2-nitrobenzoic acid ≥98.0%
4-Fluoro-2-nitrobenzoic acid

CAS Number: 394-01-4
MDL Number: MFCD00024300
Molecular Formula: C7H4FNO4
Molecular Weight: 185.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 150

Formula: C₇H₄FNO₄
MW: 185.11
Melting Pt: 150 °C
MDL Number: MFCD00024300
CAS Number: 394-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5eb828949a99f25fddd63d57a5d06619b36e4a11&vid=250737f719f98c62f16be091260a4665168b5c1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR