Flupirtine maleate salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0736-100MG F0736-1G
TCF0736-100MGEA 319.16 CAD
TCF0736-100MG TCF0736-1G
Flupirtine maleate salt ≥98.0% (by HPLC, titration analysis)
Flupirtine maleate salt

CAS Number: 75507-68-5
MDL Number: MFCD00941415
Molecular Formula: C15H17FN4O2
Molecular Weight: 420.40
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 177
Lambda max.: 343 nm (CH3CN)Storage Temperature: 0-10°C


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 2-Amino-3-(ethoxycarbonylamino)-6-(4-fluorobenzylamino)pyridine Maleate Salt

Formula: C₁₅H₁₇FN₄O₂
Melting Pt: 177 °C
MDL Number: MFCD00941415
CAS Number: 75507-68-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e2ef1ebb2d8641570405fad0f4f8cb8114a2d55&vid=7e2ef1ebb2d8641570405fad0f4f8cb8114a2d55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR