2-Fluoro-3-iodopyridine ≥98.0% (by GC)

Supplier: TCI America

F0733-1G F0733-5G
TCF0733-1GEA 62.88 CAD
TCF0733-1G TCF0733-5G
2-Fluoro-3-iodopyridine ≥98.0% (by GC)
2-Fluoro-3-iodopyridine

CAS Number: 113975-22-7
MDL Number: MFCD03095287
Molecular Formula: C5H3FIN
Molecular Weight: 222.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 99
Melting point (°C): 45

Formula: C₅H₃FIN
MW: 222.99 g/mol
Boiling Pt: 99 °C
Melting Pt: 45 °C
MDL Number: MFCD03095287
CAS Number: 113975-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17f36c58d22d8f568e81336746094f2fb9703e97&vid=17f36c58d22d8f568e81336746094f2fb9703e97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR