4-Fluoro-1,3-dioxolan-2-one ≥98.0% (by GC)

Supplier: TCI America

F0731-25G F0731-5G
TCF0731-25GEA 281.19 CAD
TCF0731-25G TCF0731-5G
4-Fluoro-1,3-dioxolan-2-one ≥98.0% (by GC)
4-Fluoro-1,3-dioxolan-2-one

CAS Number: 114435-02-8
MDL Number: MFCD06247543
Molecular Formula: C3H3FO3
Molecular Weight: 106.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Melting point (°C): 20
Specific Gravity (20/20): 1.50
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: 4-Fluoro-2-oxo-1,3-dioxolane, Fluoroethylene Carbonate

Formula: C₃H₃FO₃
Boiling Pt: 210 °C
Melting Pt: 20 °C
Density: 1.50 (20 °C)
MDL Number: MFCD06247543
CAS Number: 114435-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f18fd95c4319ead92550b4e0dd2bf1a71a9f0156&vid=f18fd95c4319ead92550b4e0dd2bf1a71a9f0156&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR