(±)-2-Fluoromandelic acid ≥98.0%

Supplier: TCI America

F0730-25G F0730-5G
TCF0730-25GEA 452.83 CAD
TCF0730-25G TCF0730-5G
(±)-2-Fluoromandelic acid ≥98.0%
(±)-2-Fluoromandelic acid

CAS Number: 389-31-1
MDL Number: MFCD03095382
Molecular Formula: C8H7FO3
Molecular Weight: 170.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 72
Melting point (°C): 117


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(2-Fluorophenyl)-2-hydroxyacetic Acid

Formula: C₈H₇FO₃
MW: 170.14
Boiling Pt: 72 °C
Melting Pt: 117 °C
MDL Number: MFCD03095382
CAS Number: 389-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48aed28a9a6d52ca51fca0a1a1923458a1453c7b&vid=48aed28a9a6d52ca51fca0a1a1923458a1453c7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR