Fmoc-Arg(Pbf)-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0729-25G F0729-5G
TCF0729-25GEA 321.86 CAD
TCF0729-25G TCF0729-5G
Fmoc-Arg(Pbf)-OH ≥98.0% (by HPLC, titration analysis)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine

CAS Number: 154445-77-9
MDL Number: MFCD00235804
Molecular Formula: C34H40N4O7S
Molecular Weight: 648.78
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -6 deg (C=1, DMF)
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis
Other Product Names / Synonyms: Nalpha-Fmoc-Nomega-Pbf-L-arginine, Fmoc-Arg(Pbf)-OH

Formula: C₃₄H₄₀N₄O₇S
MDL Number: MFCD00235804
CAS Number: 154445-77-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2efca92eb1498cc645f2040ca321504c4cdb2f78&vid=2efca92eb1498cc645f2040ca321504c4cdb2f78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR