Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥97.0% (by total nitrogen basis)
Supplier: TCI America
F0726-1G
F0726-5G
TCF0726-1GEA
33.18
CAD
TCF0726-1G
TCF0726-5G
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate ≥97.0% (by total nitrogen basis)
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate
CAS Number: 164298-23-1
MDL Number: MFCD02684443
Molecular Formula: C5H12F7N2P
Molecular Weight: 264.13
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 111
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: TFFH
Formula:
C₅H₁₂F₇N₂P Melting Pt: 111 °C |
MDL Number:
MFCD02684443 CAS Number: 164298-23-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...