2-(4-Fluorophenyl)thiophene ≥95.0%

Supplier: TCI America

F0724-1G F0724-5G
TCF0724-1GEA 74.42 CAD
TCF0724-1G TCF0724-5G
2-(4-Fluorophenyl)thiophene ≥95.0%
2-(4-Fluorophenyl)thiophene

CAS Number: 58861-48-6
MDL Number: MFCD06802535
Molecular Formula: C10H7FS
Molecular Weight: 178.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 53

Formula: C₁₀H₇FS
Melting Pt: 53 °C
MDL Number: MFCD06802535
CAS Number: 58861-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e96deff17536f03fdbf6410129763b0ed183e068&vid=e96deff17536f03fdbf6410129763b0ed183e068&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR