3-(4-Fluorophenyl)acrylaldehyde ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
3-(4-Fluorophenyl)-2-propenal, 4-Fluorocinnamaldehyde, 3-(4-Fluorophenyl)acrylaldehyde
CAS Number: 24654-55-5
MDL Number: MFCD03425895
Molecular Formula: C9H7FO
Molecular Weight: 150.15
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Specific Gravity (20/20): 1.16
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(4-Fluorophenyl)acrylaldehyde, 3-(4-Fluorophenyl)-2-propenal
Formula:
C₉H₇FO MW: 150.15 g/mol Boiling Pt: 95 °C Density: 1.16 (20 °C) |
MDL Number:
MFCD03425895 CAS Number: 24654-55-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...