Furan-2,5-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
F0710-25G
F0710-5G
TCF0710-25GEA
379.25
CAD
TCF0710-25G
TCF0710-5G
Furan-2,5-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)
Furan-2,5-dicarboxylic acid
CAS Number: 3238-40-2
MDL Number: MFCD00016582
Molecular Formula: C6H4O5
Molecular Weight: 156.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 320
Formula:
C₆H₄O₅ Melting Pt: 320 °C |
MDL Number:
MFCD00016582 CAS Number: 3238-40-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...