Formic acid ≥98.0% (by GC) for LC-MS
Supplier: TCI America
|
|
Synonyms:
Aminic Acid, Methanoic acid
F0654-25ML
F0654-5ML
TCF0654-25MLEA
145.17
CAD
TCF0654-25ML
TCF0654-5ML
Formic acid ≥98.0% (by GC) for LC-MS
Formic acid
CAS Number: 64-18-6
MDL Number: MFCD00003297
Molecular Formula: CH2O2
Molecular Weight: 46.03
Purity/Analysis Method:
Formula:
HCO₂H Boiling Pt: 100 °C Melting Pt: 8 °C Density: 1.22 (20 °C) Flash Pt: 57 °C |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 UN: 1779 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...