2-Fluoro-5-nitrobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

F0618-1G F0618-5G
TCF0618-1GEA 42.66 CAD
TCF0618-1G TCF0618-5G
2-Fluoro-5-nitrobenzaldehyde ≥98.0% (by GC)
2-Fluoro-5-nitrobenzaldehyde

CAS Number: 27996-87-8
MDL Number: MFCD00042298
Molecular Formula: C7H4FNO3
Molecular Weight: 169.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 59

Formula: C₇H₄FNO₃
Melting Pt: 59 °C
MDL Number: MFCD00042298
CAS Number: 27996-87-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=173963abc51b4d7f7ca461d726c29e820d5b33e1&vid=173963abc51b4d7f7ca461d726c29e820d5b33e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR