5-Fluoro-2-nitrobenzotrifluoride ≥98.0%

Supplier: TCI America

Synonyms: 5-Fluoro-2-nitrobenzotrifluoride, α,α,α,5-Tetrafluoro-2-nitrotoluene, 1-Fluoro-4-nitro-3-(trifluoromethyl)benzene

F0497-25G F0497-5G
TCF0497-25GEA 161.95 CAD
TCF0497-25G TCF0497-5G
5-Fluoro-2-nitrobenzotrifluoride ≥98.0%
5-Fluoro-2-nitrobenzotrifluoride

CAS Number: 393-09-9
MDL Number: MFCD00039741
Molecular Formula: C7H3F4NO2
Molecular Weight: 209.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 89
Specific Gravity (20/20): 1.53


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Nitro-alpha,alpha,alpha,5-tetrafluorotoluene, 4-Fluoro-1-nitro-2-(trifluoromethyl)benzene

Formula: C₇H₃F₄NO₂
MW: 209.1
Boiling Pt: 199 °C
Melting Pt: 23.5
Density: 1.53 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00039741
CAS Number: 393-09-9
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eadb7615dd768d50f6f8e09ad2b21314c1f192c&vid=6eadb7615dd768d50f6f8e09ad2b21314c1f192c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR