4-Fluorophenethyl bromide ≥95.0% (by GC)
Supplier: TCI America
F0431-1G
F0431-5G
TCF0431-1GEA
56.88
CAD
TCF0431-1G
TCF0431-5G
4-Fluorophenethyl bromide ≥95.0% (by GC)
4-Fluorophenethyl bromide
CAS Number: 332-42-3
MDL Number: MFCD03844794
Molecular Formula: C8H8BrF
Molecular Weight: 203.05
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 93
Specific Gravity (20/20): 1.45
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(2-Bromoethyl)-4-fluorobenzene, 4-Fluorophenethyl Bromide
Formula:
C₈H₈BrF Boiling Pt: 102 °C Density: 1.45 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD03844794 CAS Number: 332-42-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...