3-Fluoro-DL-phenylalanine 98,0
Supplier: TCI America
Synonyms:
2-Amino-3-(3-fluorophenyl)propionic acid, H-DL-Phe(3-F)-OH
F0169-1G
F0169-5G
TCF0169-1GEA
85.95
CAD
TCF0169-1G
TCF0169-5G
3-Fluoro-DL-phenylalanine 98,0
3-Fluoro-DL-phenylalanine
CAS Number: 456-88-2
MDL Number: MFCD00004273
Molecular Formula: C9H10FNO2
Molecular Weight: 183.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Amino-3-(3-fluorophenyl)propionic Acid, H-DL-Phe(3-F)-OH
Formula:
C₉H₁₀FNO₂ MW: 183.2 g/mol |
MDL Number:
MFCD00004273 CAS Number: 456-88-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...