3-Fluorophenol ≥98.0% (by GC)
Supplier: TCI America
F0159-100ML
F0159-25ML
TCF0159-100MLEA
442.4
CAD
TCF0159-100ML
TCF0159-25ML
3-Fluorophenol ≥98.0% (by GC)
3-Fluorophenol
CAS Number: 372-20-3
MDL Number: MFCD00002254
Molecular Formula: C6H5FO
Molecular Weight: 112.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 71
Freezing point (°C): 10
Specific Gravity (20/20): 1.22
Formula:
C₆H₅FO Boiling Pt: 178 °C Density: 1.22 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00002254 CAS Number: 372-20-3 UN: 2810 ADR: 6.1,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...