9-Fluorenone oxime ≥97.0% (by GC, total nitrogen)

Supplier: TCI America

F0023-1G F0023-5G
TCF0023-1GEA 26.86 CAD
TCF0023-1G TCF0023-5G
9-Fluorenone oxime ≥97.0% (by GC, total nitrogen)
9-Fluorenone oxime

CAS Number: 2157-52-0
MDL Number: MFCD00016356
Molecular Formula: C13H9NO
Molecular Weight: 195.22
Purity/Analysis Method: >97.0% (GC,N)
Form: Crystal
Melting point (°C): 194

Formula: C₁₃H₉NO
Melting Pt: 194 °C
MDL Number: MFCD00016356
CAS Number: 2157-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31e3e05905a80184bdb5d09107ad0a0b82fb10eb&vid=31e3e05905a80184bdb5d09107ad0a0b82fb10eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR