6-Ethoxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(6-Ethoxynaphthalen-2-yl)boronic acid
E1107-1G
E1107-5G
TCE1107-1GEA
60.04
CAD
TCE1107-1G
TCE1107-5G
6-Ethoxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
6-Ethoxy-2-naphthaleneboronic acid
CAS Number: 352525-98-5
MDL Number: MFCD03427009
Molecular Formula: C12H13BO3
Molecular Weight: 216.04
Form: Crystal
Color: White
Melting point (°C): 255
Formula:
C₁₂H₁₃BO₃ Melting Pt: 255 °C |
MDL Number:
MFCD03427009 CAS Number: 352525-98-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...