Ethyl-2-amino-4-methylthiophene-3-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-amino-4-methylthiophene-3-carboxylate

E1103-1G E1103-5G
TCE1103-1GEA 52.14 CAD
TCE1103-1G TCE1103-5G
Ethyl-2-amino-4-methylthiophene-3-carboxylate ≥98.0%
Ethyl 2-amino-4-methylthiophene-3-carboxylate

CAS Number: 43088-42-2
MDL Number: MFCD00051669
Molecular Formula: C8H11NO2S
Molecular Weight: 185.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 81


Other Product Names / Synonyms: 2-Amino-4-methylthiophene-3-carboxylic Acid Ethyl Ester

Formula: C₈H₁₁NO₂S
Melting Pt: 81 °C
MDL Number: MFCD00051669
CAS Number: 43088-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fff7bcd38d42d9ef140651fcaee91ff0ea8d9686&vid=fff7bcd38d42d9ef140651fcaee91ff0ea8d9686&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR