Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate ≥96.0%

Supplier: TCI America

Synonyms: Ethyl ethoxymethylene-3-oxo-4,4,4-trifluorobutyrate, 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylic Acid Ethyl Ester, Ethyl 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylate, 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyric Acid Ethyl Ester

E1087-25G E1087-5G
TCE1087-25GEA 261.17 CAD
TCE1087-25G TCE1087-5G
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate ≥96.0%
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate

CAS Number: 571-55-1
MDL Number: MFCD02677683
Molecular Formula: C9H11F3O4
Molecular Weight: 240.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 104
Specific Gravity (20/20): 1.24
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Ethoxymethylene)-4,4,4-trifluoro-3-oxobutyric Acid Ethyl Ester, Ethyl 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylate, 3-Ethoxy-2-(2,2,2-trifluoroacetyl)acrylic Acid Ethyl Ester

Formula: C₉H₁₁F₃O₄
MW: 240.18 g/mol
Boiling Pt: 120 °C
Density: 1.24 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD02677683
CAS Number: 571-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd44df465cb2571b7cb73d307992cb7fccbdfde1&vid=fd44df465cb2571b7cb73d307992cb7fccbdfde1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR