Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

E1079-1G E1079-5G
TCE1079-1GEA 52.93 CAD
TCE1079-1G TCE1079-5G
Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate ≥97.0% (by HPLC, titration analysis)
Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate

CAS Number: 641569-96-2
MDL Number: MFCD11100711
Molecular Formula: C11H15N3O2
Molecular Weight: 284.27
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 199


Other Product Names / Synonyms: 3-Carbamimidoylamino-4-methylbenzoic Acid Ethyl Ester Nitrate, Ethyl 3-Carbamimidoylamino-p-toluate Nitrate, 3-Carbamimidoylamino-p-toluic Acid Ethyl Ester Nitrate

Formula: C₁₁H₁₅N₃O₂
Melting Pt: 199 °C
MDL Number: MFCD11100711
CAS Number: 641569-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc19f850a29ce0ed2eaec41b70de4ce5aa666ea6&vid=cc19f850a29ce0ed2eaec41b70de4ce5aa666ea6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR