Ethyl-2,4-dimethylpyrrole-3-carboxylate ≥98.0% (by GC)

Supplier: TCI America

E1075-1G E1075-5G
TCE1075-1GEA 69.52 CAD
TCE1075-1G TCE1075-5G
Ethyl-2,4-dimethylpyrrole-3-carboxylate ≥98.0% (by GC)
Ethyl 2,4-dimethylpyrrole-3-carboxylate

CAS Number: 2199-51-1
MDL Number: MFCD01365811
Molecular Formula: C9H13NO2
Molecular Weight: 167.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 76
Lambda max.: 258 nm (EtOH)


Other Product Names / Synonyms: 2,4-Dimethylpyrrole-3-carboxylic Acid Ethyl Ester

Formula: C₉H₁₃NO₂
Boiling Pt: 162 °C
Melting Pt: 76 °C
MDL Number: MFCD01365811
CAS Number: 2199-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e24f2ceaae50633a8779928dabfabdb37ca000cb&vid=e24f2ceaae50633a8779928dabfabdb37ca000cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR