5-Ethynyl-2'-deoxyuridine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2′-Deoxy-5-ethynyluridine, 5-Ethynyl-2'-dU, EdU

E1057-200MG E1057-50MG
TCE1057-200MGEA 465.77 CAD
TCE1057-200MG TCE1057-50MG
5-Ethynyl-2'-deoxyuridine ≥98.0% (by HPLC, titration analysis)
5-Ethynyl-2'-deoxyuridine

CAS Number: 61135-33-9
MDL Number: MFCD01675687
Molecular Formula: C11H12N2O5
Molecular Weight: 252.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 199
Specific rotation [a]20/D: 23 deg (C=0.7, MeOH)


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Bioscience, Nucleosides and Their Analogs
Other Product Names / Synonyms: EdU

Formula: C₁₁H₁₂N₂O₅
MW: 252.23 g/mol
Melting Pt: 199 °C
MDL Number: MFCD01675687
CAS Number: 61135-33-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48bb65d3f8f48115a0308b8b08085d50feb9858f&vid=48bb65d3f8f48115a0308b8b08085d50feb9858f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR