Imidazole-4-carboxylic acid ethyl ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl imidazole-4-carboxylate

E1042-1G E1042-5G
TCE1042-1GEA 44.24 CAD
TCE1042-1G TCE1042-5G
Imidazole-4-carboxylic acid ethyl ester ≥98.0% (by GC, titration analysis)
Imidazole-4-carboxylic acid ethyl ester

CAS Number: 23785-21-9
MDL Number: MFCD07363812
Molecular Formula: C6H8N2O2
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 159
Lambda max.: 234 nm (CH3CN)


Other Product Names / Synonyms: 4-Imidazolecarboxylic Acid Ethyl Ester

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Melting Pt: 159 °C
MDL Number: MFCD07363812
CAS Number: 23785-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34050a89b6f95fca68ed48c34e6c2aee875407e0&vid=dc10d06d99aa351c0293ebd71f1cc74df09ccd10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR