(4-Hydroxyphenyl)acetic acid ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Ethyl (4-hydroxyphenyl)acetate, Ethyl 2-(4-hydroxyphenyl)acetate

E1034-25G E1034-5G
TCE1034-25GEA 270.18 CAD
TCE1034-25G TCE1034-5G
(4-Hydroxyphenyl)acetic acid ethyl ester ≥98.0%
(4-Hydroxyphenyl)acetic acid ethyl ester

CAS Number: 17138-28-2
MDL Number: MFCD00016491
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 145
Freezing point (°C): 32


Other Product Names / Synonyms: 4-Hydroxyphenylacetic Acid Ethyl Ester

Formula: C₁₀H₁₂O₃
MW: 180.2
Boiling Pt: 145 °C
MDL Number: MFCD00016491
CAS Number: 17138-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef32fe3381b6ce2ba8902f22d19a48173f054acf&vid=ef32fe3381b6ce2ba8902f22d19a48173f054acf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR