Ethyl pyrrole-2-carboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 1H-pyrrole-2-carboxylate

E1030-25G E1030-5G
TCE1030-25GEA 388.68 CAD
TCE1030-25G TCE1030-5G
Ethyl pyrrole-2-carboxylate ≥98.0%
Ethyl pyrrole-2-carboxylate

CAS Number: 2199-43-1
MDL Number: MFCD00817049
Molecular Formula: C7H9NO2
Molecular Weight: 139.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 235
Melting point (°C): 41


Other Product Names / Synonyms: Pyrrole-2-carboxylic Acid Ethyl Ester

Formula: C₇H₉NO₂
MW: 139.16 g/mol
Boiling Pt: 235 °C
Melting Pt: 41 °C
MDL Number: MFCD00817049
CAS Number: 2199-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de6b2c4c070b9aa17a1af0bc8a7ce7181297da72&vid=de6b2c4c070b9aa17a1af0bc8a7ce7181297da72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR