Ethyl-4-phenylbutyrate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl 4-phenylbutanoate, 4-Phenylbutyric Acid Ethyl Ester
E1027-5G
E1027-1G
TCE1027-5GEA
102.54
CAD
TCE1027-5G
TCE1027-1G
Ethyl-4-phenylbutyrate ≥98.0%
Ethyl-4-phenylbutyrate
CAS Number: 10031-93-3
MDL Number: MFCD00026923
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 4-Phenylbutyric Acid Ethyl Ester
Formula:
C₁₂H₁₆O₂ MW: 192.26 g/mol Boiling Pt: 100 °C Density: 1.00 (20 °C) |
MDL Number:
MFCD00026923 CAS Number: 10031-93-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...