Erdosteine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
E1026-1G
E1026-5G
TCE1026-1GEA
82.16
CAD
TCE1026-1G
TCE1026-5G
Erdosteine ≥98.0% (by GC, titration analysis)
Erdosteine
CAS Number: 84611-23-4
MDL Number: MFCD00867664
Molecular Formula: C8H11NO4S2
Molecular Weight: 249.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 162
Lambda max.: 236 nm (H2O)
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: N-(Tetrahydro-2-oxo-3-thienyl)-3-thiapentanedioic Acid Monoamide
Formula:
C₈H₁₁NO₄S₂ Melting Pt: 162 °C |
MDL Number:
MFCD00867664 CAS Number: 84611-23-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...