Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 98,0

Supplier: TCI America

Synonyms: Ethyl 3-(trifluoromethyl)pyrazole-4-carboxylate, 3-(Trifluoromethyl)pyrazole-4-carboxylic Acid Ethyl Ester, Ethyl 3-trifluoromethylpyrazole-4-carboxylate

E1024-1G E1024-5G
TCE1024-1GEA 94.8 CAD
TCE1024-1G TCE1024-5G
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 98,0
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate

CAS Number: 155377-19-8
MDL Number: MFCD00052083
Molecular Formula: C7H7F3N2O2
Molecular Weight: 208.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 145


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Trifluoromethyl)pyrazole-4-carboxylic Acid Ethyl Ester

Formula: C₇H₇F₃N₂O₂
MW: 208.14 g/mol
Melting Pt: 145 °C
MDL Number: MFCD00052083
CAS Number: 155377-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4f544add83a3e0372d743a5de057e1a1d97e90e&vid=d4f544add83a3e0372d743a5de057e1a1d97e90e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR