Ethyl-3-acetoxyhexanoate ≥98.0%

Supplier: TCI America

Synonyms: 3-Acetoxyhexanoic Acid Ethyl Ester

E1014-25G E1014-5G
TCE1014-25GEA 395 CAD
TCE1014-25G TCE1014-5G
Ethyl-3-acetoxyhexanoate ≥98.0%
Ethyl-3-acetoxyhexanoate

CAS Number: 21188-61-4
MDL Number: MFCD00792508
Molecular Formula: C10H18O4
Molecular Weight: 202.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 3-Acetoxyhexanoic Acid Ethyl Ester

Formula: C₁₀H₁₈O₄
Boiling Pt: 116 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00792508
CAS Number: 21188-61-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d955479d32594e2cc5b6be93e82b5ff99de2fe3&vid=1d955479d32594e2cc5b6be93e82b5ff99de2fe3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR